2,5-Di-tert-butylhydroquinone 98%

Supplier: Ambeed
Danger

A348334-100G A348334-100MG A348334-25G A348334-1000G
76876-254EA 16 USD
76876-254 76876-256 76876-258 77883-953
2,5-Di-tert-butylhydroquinone 98%
2,5-Di-tert-butylhydroquinone

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₂
MDL Number: MFCD00008825
CAS Number: 88-58-4
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR