tert-Butyl 3,4-dihydroquinoline-1(2H)-carboxylate 98%

Supplier: Ambeed
Warning

A347402-10G A347402-1G
76876-244EA 221.73 USD
76876-244 76876-246
tert-Butyl 3,4-dihydroquinoline-1(2H)-carboxylate 98%
tert-Butyl 3,4-dihydroquinoline-1(2H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MW: 233.30 g/mol
MDL Number: MFCD19684135
CAS Number: 123387-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR