(3-Bromo-5-isobutoxyphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-5-isobutoxyphenylboronic acid

A346133-1G A346133-5G
76876-228EA 18.04 USD
76876-228 76876-230
(3-Bromo-5-isobutoxyphenyl)boronic acid 98%
(3-Bromo-5-isobutoxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BBrO₃
MW: 272.93 g/mol
MDL Number: MFCD17214245
CAS Number: 918904-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR