4-Chloro-6-hydrazinopyrimidine 98%

Supplier: Ambeed
Danger

A345220-1G A345220-250MG
76876-218EA 22 USD
76876-218 76876-220
4-Chloro-6-hydrazinopyrimidine 98%
4-Chloro-6-hydrazinopyrimidine

Precaución: for Research Use Only

Formula: C₄H₅ClN₄
MW: 144.56 g/mol
MDL Number: MFCD01646095
CAS Number: 5767-35-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR