(1R,2R)-Boc-Acpc 95%

Supplier: Ambeed
Warning

A344509-250MG A344509-1G A344509-100MG
76876-216EA 54 USD
76876-216 77672-752 77883-948
(1R,2R)-Boc-Acpc 95%
(1R,2R)-Boc-Acpc

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD04974223
CAS Number: 245115-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR