2-Fluoro-3-isobutoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Fluoro-3-isobutoxyphenyl)boronic acid

A343734-100MG A343734-1G A343734-250MG
76876-206EA 37.14 USD
76876-206 76876-208 76876-210
2-Fluoro-3-isobutoxyphenylboronic acid 98%
2-Fluoro-3-isobutoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
MW: 212.03 g/mol
MDL Number: MFCD12546465
CAS Number: 1217500-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR