(2R,3R,4S,5R)-2-(4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 95%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-7-deazapurine-β-D-riboside

A342436-1G A342436-250MG A342436-5G A342436-100MG
76876-200EA 215.37 USD
76876-200 76876-202 76876-204 77672-744
(2R,3R,4S,5R)-2-(4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol 95%
(2R,3R,4S,5R)-2-(4-Chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClN₃O₄
MW: 285.68 g/mol
MDL Number: MFCD03425528
CAS Number: 16754-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR