tert-Butyl 4-(2-nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A341309-1G A341309-250MG A341309-5G
76876-188EA 152 USD
76876-188 76876-190 76876-192
tert-Butyl 4-(2-nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine-1-carboxylate 95%
tert-Butyl 4-(2-nitro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₂₁H₃₂BN₃O₆
MDL Number: MFCD13195254
CAS Number: 940284-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR