4,5-Dimethyl-1,3-thiazole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A339964-100MG A339964-250MG A339964-5G A339964-1G
76876-168EA 30 USD
76876-168 76876-170 76876-172 77672-730
4,5-Dimethyl-1,3-thiazole-2-carboxylic acid 98%
4,5-Dimethyl-1,3-thiazole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MDL Number: MFCD11934399
CAS Number: 79312-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR