Methyl 2,4-dihydroxy-5-nitrobenzoate 95%

Supplier: Ambeed
Warning

A339925-100MG A339925-1G A339925-250MG A339925-5G
76876-162EA 5 USD
76876-162 76876-164 76876-166 77612-950
Methyl 2,4-dihydroxy-5-nitrobenzoate 95%
Methyl 2,4-dihydroxy-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇NO₆
MDL Number: MFCD10699564
CAS Number: 271261-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR