Ethyl 5-fluoro-3-iodo-1H-indole-2-carboxylate 96%

Supplier: Ambeed
Warning

A339752-1G A339752-250MG
76876-158EA 149.59 USD
76876-158 76876-160
Ethyl 5-fluoro-3-iodo-1H-indole-2-carboxylate 96%
Ethyl 5-fluoro-3-iodo-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉FINO₂
MW: 333.1 g/mol
MDL Number: MFCD13184347
CAS Number: 167631-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR