Ethyl 7-bromo-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate 98%

Supplier: Ambeed
Warning

A337958-1G A337958-250MG A337958-5G
76876-128EA 89.12 USD
76876-128 76876-130 76876-132
Ethyl 7-bromo-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate 98%
Ethyl 7-bromo-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂O₃
MW: 297.10 g/mol
MDL Number: MFCD12546518
CAS Number: 16867-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR