1-(2-Chloroethyl)-4-methylpiperazine dihydrochloride 95%

Supplier: Ambeed
Warning

A337422-10G A337422-1G A337422-250MG A337422-25G A337422-5G
76876-114EA 211 USD
76876-114 76876-116 76876-118 76876-120 76876-122
1-(2-Chloroethyl)-4-methylpiperazine dihydrochloride 95%
1-(2-Chloroethyl)-4-methylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₇Cl₃N₂
MW: 235.58 g/mol
MDL Number: MFCD17926367
CAS Number: 5753-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR