Butane-1,2,4-tricarboxylic acid 97%

Supplier: Ambeed
Warning

A336460-1G A336460-25G A336460-5G
76876-092EA 20.16 USD
76876-092 76876-094 76876-096
Butane-1,2,4-tricarboxylic acid 97%
Butane-1,2,4-tricarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₆
MW: 190.15 g/mol
MDL Number: MFCD00152304
CAS Number: 923-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR