2-Cyclopropyl-2-oxoacetic acid 95%

Supplier: Ambeed
Warning

A334824-1G A334824-250MG
76876-064EA 82.76 USD
76876-064 76876-066
2-Cyclopropyl-2-oxoacetic acid 95%
2-Cyclopropyl-2-oxoacetic acid

Precaución: for Research Use Only

Formula: C₅H₆O₃
MW: 114.09 g/mol
MDL Number: MFCD12024468
CAS Number: 13885-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR