4-Trifluoromethyl-trans-beta-nitrostyrene 97%

Supplier: Ambeed
Warning

A334458-1G A334458-250MG
76876-046EA 133.68 USD
76876-046 76876-048
4-Trifluoromethyl-trans-beta-nitrostyrene 97%
4-Trifluoromethyl-trans-beta-nitrostyrene

Precaución: for Research Use Only

Formula: C₉H₆F₃NO₂
MW: 217.14 g/mol
MDL Number: MFCD00192359
CAS Number: 93628-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR