4-(2-Bromophenylcarbamoyl)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-((2-Bromophenyl)carbamoyl)phenyl)boronic acid

A333724-1G A333724-5G
76876-040EA 13 USD
76876-040 76876-042
4-(2-Bromophenylcarbamoyl)phenylboronic acid 95%
4-(2-Bromophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BBrNO₃
MDL Number: MFCD09027202
CAS Number: 874288-01-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR