3-(Bromoacetyl)-5-chloro-2-thiophenesulfonamide 95%

Supplier: Ambeed
Danger

Synonyms: 3-(2-Bromoacetyl)-5-chlorothiophene-2-sulphonamide

A332107-10G A332107-1G
76876-028EA 125.19 USD
76876-028 76876-030
3-(Bromoacetyl)-5-chloro-2-thiophenesulfonamide 95%
3-(Bromoacetyl)-5-chloro-2-thiophenesulfonamide

Precaución: for Research Use Only

Formula: C₆H₅BrClNO₃S₂
MW: 318.596 g/mol
MDL Number: MFCD09033310
CAS Number: 160982-11-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR