7-Bromo-2-methyl-1H-indene 97%

Supplier: Ambeed
Warning

A331472-10G A331472-1G A331472-5G
76876-016EA 77 USD
76876-016 76876-018 76876-020
7-Bromo-2-methyl-1H-indene 97%
7-Bromo-2-methyl-1H-indene

Precaución: for Research Use Only

Formula: C₁₀H₉Br
MDL Number: MFCD20524977
CAS Number: 880652-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR