(R)-Piperidin-3-ol 97%

Supplier: Ambeed
Danger

A330550-100MG A330550-1G A330550-250MG A330550-5G
76875-992EA 31.83 USD
76875-992 76875-994 76875-996 76875-998
(R)-Piperidin-3-ol 97%
(R)-Piperidin-3-ol

Precaución: for Research Use Only

Formula: C₅H₁₁NO
MW: 101.14 g/mol
MDL Number: MFCD09832542
CAS Number: 62414-68-0
UN: 3263
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR