1,2-Dibromo-4-(trifluoromethoxy)benzene 98+%

Supplier: Ambeed
Warning

A330420-1G A330420-25G A330420-5G
76875-982EA 5 USD
76875-982 77883-903 77883-904
1,2-Dibromo-4-(trifluoromethoxy)benzene 98+%
1,2-Dibromo-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₃Br₂F₃O
MDL Number: MFCD23099535
CAS Number: 1682-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR