Methyl(2,2,2-trifluoroethyl)amine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-2,2,2-trifluoroethylamine Hydrochloride

A328937-100MG A328937-250MG A328937-5G A328937-1G
76875-976EA 62.6 USD
76875-976 76875-978 76875-980 77593-944
Methyl(2,2,2-trifluoroethyl)amine hydrochloride 95%
Methyl(2,2,2-trifluoroethyl)amine hydrochloride

Precaución: for Research Use Only

Formula: C₃H₇ClF₃N
MW: 149.54 g/mol
MDL Number: MFCD08448126
CAS Number: 2730-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR