4-(4-Bromophenyl)cyclohexanone 95%

Supplier: Ambeed
Warning

A328861-100MG A328861-250MG A328861-5G
76875-974EA 41 USD
76875-974 77672-688 77883-897
4-(4-Bromophenyl)cyclohexanone 95%
4-(4-Bromophenyl)cyclohexanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrO
MDL Number: MFCD11111144
CAS Number: 84892-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR