(1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)methanol 98%
Supplier: Ambeed
|
Warning
|
A326810-100MG
A326810-1G
A326810-250MG
A326810-5G
76875-956EA
51
USD
76875-956
76875-958
76875-960
76875-962
(1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)methanol 98%
(1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)methanol
Precaución: for Research Use Only
Formula:
C₁₆H₂₃BO₃ |
MDL Number:
MFCD18383801 CAS Number: 1220219-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...