4-Bromo-6-chloroquinoline 97%

Supplier: Ambeed
Danger

A324281-1G A324281-250MG A324281-5G
76875-926EA 87 USD
76875-926 76875-928 76875-930
4-Bromo-6-chloroquinoline 97%
4-Bromo-6-chloroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MDL Number: MFCD11505112
CAS Number: 1070879-30-9
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR