3-(4-Bromo-3-fluorophenyl)acrylic acid 98%

Supplier: Ambeed
Warning

A324248-100MG A324248-1G A324248-250MG
76875-916EA 45.62 USD
76875-916 76875-918 76875-920
3-(4-Bromo-3-fluorophenyl)acrylic acid 98%
3-(4-Bromo-3-fluorophenyl)acrylic acid

Precaución: for Research Use Only

Formula: C₉H₆BrFO₂
MW: 245.05 g/mol
MDL Number: MFCD10000928
CAS Number: 923266-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR