(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl) ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (S,S)-DACH-Naphthyl Trost Ligand, N,N'-((1S,2S)-Cyclohexane-1,2-diyl)bis(2-(diphenylphosphino)-1-naphthamide)

A323354-100MG A323354-1G A323354-250MG A323354-5G
76875-896EA 25.47 USD
76875-896 76875-898 76875-900 76875-902
(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl) ≥98%, ee 99%
(1S,2S)-(-)-1,2-Diaminocyclohexane-N,N'-bis(2-diphenylphosphino-1-naphthoyl)

Precaución: for Research Use Only

Formula: C₅₂H₄₄N₂O₂P₂
MW: 790.8800 g/mol
MDL Number: MFCD02684552
CAS Number: 205495-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR