N-(4-Fluorophenyl) 4-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-[N-(4-Fluorophenyl)sulphamoyl]benzeneboronic acid, (4-(N-(4-Fluorophenyl)sulphamoyl)phenyl)boronic acid

A323080-1G A323080-250MG A323080-5G
76875-890EA 79.57 USD
76875-890 76875-892 76875-894
N-(4-Fluorophenyl) 4-boronobenzenesulfonamide 98%
N-(4-Fluorophenyl) 4-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₂H₁₁BFNO₄S
MW: 295.09 g/mol
MDL Number: MFCD09800889
CAS Number: 957121-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR