2-Fluoro-3-methoxybenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2-fluoro-3-methoxybenzene

A322617-1G A322617-250MG A322617-5G
76875-878EA 44 USD
76875-878 76875-880 76875-882
2-Fluoro-3-methoxybenzyl bromide 98%
2-Fluoro-3-methoxybenzyl bromide

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MDL Number: MFCD09743597
CAS Number: 447463-56-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR