4-(Cyclopropylcarbamoyl)-3-fluorophenylboronic acid 98%

Supplier: Ambeed
Danger

A322401-1G A322401-250MG A322401-5G
76875-872EA 128 USD
76875-872 76875-874 76875-876
4-(Cyclopropylcarbamoyl)-3-fluorophenylboronic acid 98%
4-(Cyclopropylcarbamoyl)-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BFNO₃
MDL Number: MFCD09972092
CAS Number: 874289-20-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR