(S)-b-Amino-4-chloro-benzeneethanol 98%

Supplier: Ambeed
Warning

A322270-1G A322270-250MG A322270-100MG
76875-868EA 85 USD
76875-868 76875-870 77883-871
(S)-b-Amino-4-chloro-benzeneethanol 98%
(S)-b-Amino-4-chloro-benzeneethanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO
MDL Number: MFCD09253716
CAS Number: 191109-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR