2-(4-Chlorobenzoyl)benzoic acid 98%

Supplier: Ambeed
Warning

A322264-1000G A322264-100G A322264-10G A322264-25G
76875-860EA 125.19 USD
76875-860 76875-862 76875-864 76875-866
2-(4-Chlorobenzoyl)benzoic acid 98%
2-(4-Chlorobenzoyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₉ClO₃
MW: 260.68 g/mol
MDL Number: MFCD00002474
CAS Number: 85-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR