N-Boc-4-methyl-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-p-Me-Phe-OH, Boc-Phe(4-Me)-OH

A321543-10G A321543-250MG A321543-25G A321543-5G
76875-850EA 118 USD
76875-850 76875-852 76875-854 77883-870
N-Boc-4-methyl-L-phenylalanine 97%
N-Boc-4-methyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
MDL Number: MFCD01317027
CAS Number: 80102-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR