(3-(N-Isopropyl-N-(4-methoxybenzyl)sulfamoyl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A319751-100MG A319751-1G A319751-250MG
76875-844EA 36.08 USD
76875-844 76875-846 76875-848
(3-(N-Isopropyl-N-(4-methoxybenzyl)sulfamoyl)phenyl)boronic acid 96%
(3-(N-Isopropyl-N-(4-methoxybenzyl)sulfamoyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₂BNO₅S
MW: 363.24 g/mol
MDL Number: MFCD12756420
CAS Number: 1217501-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR