Methyl 2-amino-3-chloro-5-(trifluoromethoxy)benzoate 98+%

Supplier: Ambeed
Warning

A319717-100MG A319717-250MG
76875-840EA 65.78 USD
76875-840 76875-842
Methyl 2-amino-3-chloro-5-(trifluoromethoxy)benzoate 98+%
Methyl 2-amino-3-chloro-5-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₇ClF₃NO₃
MW: 269.61 g/mol
MDL Number: MFCD09835202
CAS Number: 1003708-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR