3-(Boc-amino)cyclohexanecarboxylic acid 97%

Supplier: Ambeed
Warning

A319500-10G A319500-250MG A319500-5G A319500-1G A319500-25G
76875-834EA 125 USD
76875-834 76875-836 76875-838 77593-892 77883-868
3-(Boc-amino)cyclohexanecarboxylic acid 97%
3-(Boc-amino)cyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MDL Number: MFCD03453252
CAS Number: 334932-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR