4,6-Dibromodibenzothiophene 97%

Supplier: Ambeed
Warning

A319288-1G A319288-250MG A319288-5G A319288-25G
76875-828EA 27 USD
76875-828 76875-830 76875-832 77672-642
4,6-Dibromodibenzothiophene 97%
4,6-Dibromodibenzothiophene

Precaución: for Research Use Only

Formula: C₁₂H₆Br₂S
MDL Number: MFCD18451473
CAS Number: 669773-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR