1-Boc-5-methoxyindole-3-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

A315682-100MG A315682-1G A315682-250MG A315682-5G
76875-794EA 36 USD
76875-794 76875-796 76875-798 76875-800
1-Boc-5-methoxyindole-3-boronic acid pinacol ester 98%
1-Boc-5-methoxyindole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₈BNO₅
MDL Number: MFCD16295115
CAS Number: 1256359-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR