1-Methyl-1H-indazole-4-boronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole

A312044-100MG A312044-1G A312044-250MG
76875-772EA 10 USD
76875-772 76875-774 76875-776
1-Methyl-1H-indazole-4-boronic acid pinacol ester 96%
1-Methyl-1H-indazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD10700158
CAS Number: 885698-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR