(5-Methylisoxazol-3-yl)methanamine hydrochloride 97%

Supplier: Ambeed
Warning

A311062-100MG A311062-1G A311062-250MG
76875-756EA 43.5 USD
76875-756 76875-758 77612-788
(5-Methylisoxazol-3-yl)methanamine hydrochloride 97%
(5-Methylisoxazol-3-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉ClN₂O
MW: 148.59 g/mol
MDL Number: MFCD00464228
CAS Number: 1050590-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR