2-(5-Bromopyridin-2-yl)propan-2-ol 97%

Supplier: Ambeed
Warning

A310110-100MG A310110-1G A310110-250MG
76875-752EA 32.89 USD
76875-752 76875-754 77672-580
2-(5-Bromopyridin-2-yl)propan-2-ol 97%
2-(5-Bromopyridin-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
MDL Number: MFCD11865230
CAS Number: 290307-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR