(2,6-Difluoro-4-formylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-4-formylphenylboronic acid

A309445-1G A309445-250MG A309445-5G
76875-744EA 60 USD
76875-744 76875-746 76875-748
(2,6-Difluoro-4-formylphenyl)boronic acid 98%
(2,6-Difluoro-4-formylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₃
MW: 185.92 g/mol
MDL Number: MFCD10697422
CAS Number: 871125-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR