(S)-5-(Chloromethyl)oxazolidin-2-one 95%

Supplier: Ambeed
Warning

A307649-1G A307649-250MG A307649-25G A307649-5G
76875-724EA 43.5 USD
76875-724 76875-726 76875-728 76875-730
(S)-5-(Chloromethyl)oxazolidin-2-one 95%
(S)-5-(Chloromethyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₄H₆ClNO₂
MW: 135.54 g/mol
MDL Number: MFCD08706410
CAS Number: 169048-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR