1-Boc-5-cyanoindole-3-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 5-cyano-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1-carboxylate

A307542-1G A307542-250MG
76875-722EA 171 USD
76875-722 77883-838
1-Boc-5-cyanoindole-3-boronic acid pinacol ester 98%
1-Boc-5-cyanoindole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₀H₂₅BN₂O₄
MW: 368.2345 g/mol
MDL Number: MFCD15143604
CAS Number: 1185427-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR