2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(2-phenylpropan-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2H-Benzotriazol-2-yl)-6-(1-methyl-1-phenylethyl)-4-(1,1,3,3-tetramethylbutyl)phenol

A306925-100G A306925-25G A306925-5G
76875-706EA 140.04 USD
76875-706 76875-708 76875-710
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(2-phenylpropan-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol 98%
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(2-phenylpropan-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₂₉H₃₅N₃O
MW: 441.62 g/mol
MDL Number: MFCD02100741
CAS Number: 73936-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR