2,2'-Dimethyl-4,4'-bipyridine 98%

Supplier: Ambeed
Warning

A306219-100MG A306219-250MG
76875-702EA 72.15 USD
76875-702 76875-704
2,2'-Dimethyl-4,4'-bipyridine 98%
2,2'-Dimethyl-4,4'-bipyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂
MW: 184.23 g/mol
MDL Number: MFCD06637667
CAS Number: 712-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR