(4-Oxocyclohexyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Oxocyclohexyl)acetic acid

A306214-1G A306214-250MG
76875-698EA 62.6 USD
76875-698 76875-700
(4-Oxocyclohexyl)acetic acid 98%
(4-Oxocyclohexyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₁₂O₃
MW: 156.18 g/mol
MDL Number: MFCD09864369
CAS Number: 52263-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR