N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine 98%

Supplier: Ambeed
Danger

A305991-1G A305991-25G A305991-5G A305991-500G
76875-692EA 5 USD
76875-692 76875-694 76875-696 77885-812
N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine 98%
N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine

Precaución: for Research Use Only

Formula: C₁₈H₂₄N₂
MDL Number: MFCD00072248
CAS Number: 793-24-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR