(2-Methyl-4-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A304527-100MG A304527-1G A304527-250MG
76875-686EA 36.08 USD
76875-686 76875-688 76875-690
(2-Methyl-4-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid 95%
(2-Methyl-4-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₄S
MW: 269.13 g/mol
MDL Number: MFCD13195671
CAS Number: 1217501-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR