4,5-Dibromo-2-methylpyridazin-3(2H)-one 98%

Supplier: Ambeed
Warning

A303425-100MG A303425-1G A303425-250MG
76875-672EA 105.03 USD
76875-672 76875-674 76875-676
4,5-Dibromo-2-methylpyridazin-3(2H)-one 98%
4,5-Dibromo-2-methylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₅H₄Br₂N₂O
MW: 267.90 g/mol
MDL Number: MFCD00135076
CAS Number: 13645-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR